5-Methyl[2,3']bipyridinyl ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Methyl-2,3'-bipyridine
086318-1G
086318-5G
102847-974EA
860
USD
102847-974
102847-978
5-Methyl[2,3']bipyridinyl ≥97%
5-Methyl[2,3']bipyridinyl
Hazard: Irritant
Formula:
C₁₁H₁₀N₂ MW: 170.22 g/mol Melting Pt: 177…180 °C |
MDL Number:
MFCD09037514 CAS Number: 26844-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...