Methyl biphenyl-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl [1,1'-biphenyl]-2-carboxylate

086301-1G 086301-5G
102847-878EA 243.08 USD
102847-878 102847-882
Methyl biphenyl-2-carboxylate ≥97%
Methyl biphenyl-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00452250
CAS Number: 16605-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR