4-(3-Butenyl)benzoic acid ≥97%

Supplier: Matrix Scientific

086299-1G 086299-5G
102847-866EA 592.31 USD
102847-866 102847-870
4-(3-Butenyl)benzoic acid ≥97%
4-(3-Butenyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD02259926
CAS Number: 15451-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR