3-(2-Methyl-2-propenyl)benzoic acid ≥97%

Supplier: Matrix Scientific

086297-1G 086297-5G
102847-854EA 387.69 USD
102847-854 102847-858
3-(2-Methyl-2-propenyl)benzoic acid ≥97%
3-(2-Methyl-2-propenyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD02259923
CAS Number: 732249-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR