3-(2-Propenyl)benzoic acid ≥97%
Supplier: Matrix Scientific
086295-1G
086295-5G
102847-842EA
658.46
USD
102847-842
102847-846
3-(2-Propenyl)benzoic acid ≥97%
3-(2-Propenyl)benzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₀O₂ |
MDL Number:
MFCD02259917 CAS Number: 1077-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...