2-(3-Butenyl)benzoic acid ≥97%

Supplier: Matrix Scientific

086293-1G 086293-5G
102847-830EA 620 USD
102847-830 102847-834
2-(3-Butenyl)benzoic acid ≥97%
2-(3-Butenyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD02259910
CAS Number: 89730-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR