2-Methyl-3-(2-pyridyl)-1-propene ≥97%

Supplier: Matrix Scientific

086287-1G 086287-5G
102847-794EA 592.31 USD
102847-794 102847-798
2-Methyl-3-(2-pyridyl)-1-propene ≥97%
2-Methyl-3-(2-pyridyl)-1-propene

Hazard: Irritant/Refrigerate

Formula: C₉H₁₁N
MDL Number: MFCD07775126
CAS Number: 936621-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR