8-Acetoxy-2-methyl-1-octene ≥97%
Supplier: Matrix Scientific
086281-1G
086281-5G
102847-758EA
387.69
USD
102847-758
102847-762
8-Acetoxy-2-methyl-1-octene ≥97%
8-Acetoxy-2-methyl-1-octene
Hazard: Irritant
Formula:
C₁₁H₂₀O₂ |
MDL Number:
MFCD02259781 CAS Number: 731773-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...