3-(4-Methoxy-2-methylphenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086275-1G
086275-5G
102847-722EA
387.69
USD
102847-722
102847-726
3-(4-Methoxy-2-methylphenyl)-1-propene ≥97%
3-(4-Methoxy-2-methylphenyl)-1-propene
Hazard: Irritant
Formula:
C₁₁H₁₄O |
MDL Number:
MFCD07775075 CAS Number: 16964-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...