2-Bromo-4-(3,5-dimethoxyphenyl)-1-butene ≥97%
Supplier: Matrix Scientific
086264-1G
086264-5G
102847-656EA
592.31
USD
102847-656
102847-660
2-Bromo-4-(3,5-dimethoxyphenyl)-1-butene ≥97%
2-Bromo-4-(3,5-dimethoxyphenyl)-1-butene
Hazard: Irritant
Formula:
C₁₂H₁₅BrO₂ |
MDL Number:
MFCD09801200 CAS Number: 34217-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...