3-(2,6-Dimethoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086258-1G
086258-5G
102847-620EA
387.69
USD
102847-620
102847-624
3-(2,6-Dimethoxyphenyl)-2-methyl-1-propene ≥97%
3-(2,6-Dimethoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD09801190 CAS Number: 35820-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...