4-(2,5-Dimethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086254-1G 086254-5G
102847-596EA 592.31 USD
102847-596 102847-600
4-(2,5-Dimethoxyphenyl)-1-butene ≥97%
4-(2,5-Dimethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD09801180
CAS Number: 182132-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR