3-(4-n-Butylphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086247-1G 086247-5G
102847-560EA 387.69 USD
102847-560 102847-564
3-(4-n-Butylphenyl)-1-propene ≥97%
3-(4-n-Butylphenyl)-1-propene

Hazard: Irritant

Formula: C₁₃H₁₈
MDL Number: MFCD06201179
CAS Number: 842124-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR