3-(4-Ethoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086239-1G 086239-5G
102847-512EA 584.62 USD
102847-512 102847-516
3-(4-Ethoxyphenyl)-1-propene ≥97%
3-(4-Ethoxyphenyl)-1-propene

Hazard: Irritant

Formula: C₁₁H₁₄O
MDL Number: MFCD09801155
CAS Number: 3698-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR