3-[(4-Trifluoromethyl)phenyl]-1-propene ≥97%

Supplier: Matrix Scientific

086235-1G 086235-5G
102847-488EA 729.23 USD
102847-488 102847-492
3-[(4-Trifluoromethyl)phenyl]-1-propene ≥97%
3-[(4-Trifluoromethyl)phenyl]-1-propene

Hazard: Irritant

Formula: C₁₀H₉F₃
MW: 186.18 g/mol
MDL Number: MFCD09801115
CAS Number: 1813-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR