3-[(3-Trifluoromethyl)phenyl]-1-propene ≥97%

Supplier: Matrix Scientific

086233-1G 086233-5G
102847-476EA 387.69 USD
102847-476 102847-480
3-[(3-Trifluoromethyl)phenyl]-1-propene ≥97%
3-[(3-Trifluoromethyl)phenyl]-1-propene

Hazard: Irritant

Formula: C₁₀H₉F₃
MDL Number: MFCD09801107
CAS Number: 1813-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR