4-(4-Acetoxyphenyl)-1-butene ≥97%
Supplier: Matrix Scientific
086229-1G
086229-5G
102847-452EA
592.31
USD
102847-452
102847-456
4-(4-Acetoxyphenyl)-1-butene ≥97%
4-(4-Acetoxyphenyl)-1-butene
Hazard: Irritant
Formula:
C₁₂H₁₄O₂ |
MDL Number:
MFCD08064673 CAS Number: 173852-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...