4-(3-Acetoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086227-1G 086227-5G
102847-440EA 592.31 USD
102847-440 102847-444
4-(3-Acetoxyphenyl)-1-butene ≥97%
4-(3-Acetoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD08064672
CAS Number: 890097-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR