3-(2-Acetoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086225-1G
086225-5G
102847-428EA
387.69
USD
102847-428
102847-432
3-(2-Acetoxyphenyl)-2-methyl-1-propene ≥97%
3-(2-Acetoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₂H₁₄O₂ |
MDL Number:
MFCD07698661 CAS Number: 861009-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...