4-(4-Methoxyphenyl)-2-methyl-1-butene ≥97%

Supplier: Matrix Scientific

086223-1G 086223-5G
102847-416EA 592.31 USD
102847-416 102847-420
4-(4-Methoxyphenyl)-2-methyl-1-butene ≥97%
4-(4-Methoxyphenyl)-2-methyl-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD07698655
CAS Number: 18491-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR