4-(4-Methoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086221-1G 086221-5G
102847-404EA 592.31 USD
102847-404 102847-408
4-(4-Methoxyphenyl)-1-butene ≥97%
4-(4-Methoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD07698650
CAS Number: 20574-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR