3-(3-Methoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086219-1G
086219-5G
102847-392EA
387.69
USD
102847-392
102847-396
3-(3-Methoxyphenyl)-2-methyl-1-propene ≥97%
3-(3-Methoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₁H₁₄O |
MDL Number:
MFCD07698648 CAS Number: 68383-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...