3-(3-Methoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Allylanisole

086217-1G 086217-5G
102847-380EA 350.77 USD
102847-380 102847-384
3-(3-Methoxyphenyl)-1-propene ≥97%
3-(3-Methoxyphenyl)-1-propene

Hazard: Irritant-Harmful

Formula: C₁₀H₁₂O
MW: 148.21 g/mol
MDL Number: MFCD06201195
CAS Number: 24743-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR