3-(3-Fluorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086213-1G
086213-5G
102847-356EA
946.15
USD
102847-356
102847-360
3-(3-Fluorophenyl)-1-propene ≥97%
3-(3-Fluorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₉F |
MDL Number:
MFCD06201194 CAS Number: 30984-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...