3-(2-Fluorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086211-1G
086211-5G
102847-344EA
556.92
USD
102847-344
102847-348
3-(2-Fluorophenyl)-1-propene ≥97%
3-(2-Fluorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₉F |
MDL Number:
MFCD07698616 CAS Number: 56314-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...