4-(4-Carboethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086209-1G 086209-5G
102847-332EA 592.31 USD
102847-332 102847-336
4-(4-Carboethoxyphenyl)-1-butene ≥97%
4-(4-Carboethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD01319583
CAS Number: 591760-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR