4-(3-Carboethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086207-1G 086207-5G
102847-320EA 592.31 USD
102847-320 102847-324
4-(3-Carboethoxyphenyl)-1-butene ≥97%
4-(3-Carboethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD01319578
CAS Number: 731772-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR