3-(2-Carboethoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086205-1G
086205-5G
102847-308EA
387.69
USD
102847-308
102847-312
3-(2-Carboethoxyphenyl)-2-methyl-1-propene ≥97%
3-(2-Carboethoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₃H₁₆O₂ |
MDL Number:
MFCD00671912 CAS Number: 731772-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...