3-(2-Carboethoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086203-1G 086203-5G
102847-296EA 646.15 USD
102847-296 102847-300
3-(2-Carboethoxyphenyl)-1-propene ≥97%
3-(2-Carboethoxyphenyl)-1-propene

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD01319572
CAS Number: 372510-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR