3-(3-Chloro-5-fluorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086200-1G
086200-2G
086200-5G
102847-278EA
387.69
USD
102847-278
102847-280
102847-282
3-(3-Chloro-5-fluorophenyl)-1-propene ≥97%
3-(3-Chloro-5-fluorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₈ClF Boiling Pt: 67 °C (5 mmHg) |
MDL Number:
MFCD06201181 CAS Number: 842124-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...