4-(4-Methylphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(But-3-en-1-yl)toluene

086193-1G 086193-2G 086193-5G
102847-236EA 592.31 USD
102847-236 102847-238 102847-240
4-(4-Methylphenyl)-1-butene ≥97%
4-(4-Methylphenyl)-1-butene

Hazard: Irritant

Formula: C₁₁H₁₄
MW: 146.23 g/mol
MDL Number: MFCD01319558
CAS Number: 20574-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR