2-Bromo-3-(4-chlorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086191-1G
086191-2G
086191-5G
102847-224EA
946.15
USD
102847-224
102847-226
102847-228
2-Bromo-3-(4-chlorophenyl)-1-propene ≥97%
2-Bromo-3-(4-chlorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₈BrCl |
MDL Number:
MFCD00671891 CAS Number: 93507-53-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...