2-Chloro-4-(4-chlorophenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086190-1G 086190-2G 086190-5G
102847-218EA 946.15 USD
102847-218 102847-220 102847-222
2-Chloro-4-(4-chlorophenyl)-1-butene ≥97%
2-Chloro-4-(4-chlorophenyl)-1-butene

Hazard: Irritant

Formula: C₁₀H₁₀Cl₂
MDL Number: MFCD00671902
CAS Number: 731772-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR