4-(4-Chlorophenyl)-1-butene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(But-3-en-1-yl)chlorobenzene

086189-1G 086189-2G 086189-5G
102847-212EA 592.31 USD
102847-212 102847-214 102847-216
4-(4-Chlorophenyl)-1-butene ≥97%
4-(4-Chlorophenyl)-1-butene

Hazard: Irritant-Harmful/Light Sensitive

Formula: C₁₀H₁₁Cl
MW: 166.65 g/mol
MDL Number: MFCD01319548
CAS Number: 3047-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR