4-(3-Chlorophenyl)-1-butene ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(But-3-en-1-yl)-3-chlorobenzene
086187-1G
086187-2G
086187-5G
102847-200EA
592.31
USD
102847-200
102847-202
102847-204
4-(3-Chlorophenyl)-1-butene ≥97%
4-(3-Chlorophenyl)-1-butene
Hazard: Irritant
Formula:
C₁₀H₁₁Cl MW: 166.65 g/mol |
MDL Number:
MFCD01319546 CAS Number: 91426-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...