3-(3-Chlorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Allylchlorobenzene
086186-1G
086186-2G
086186-5G
102847-194EA
387.69
USD
102847-194
102847-196
102847-198
3-(3-Chlorophenyl)-1-propene ≥97%
3-(3-Chlorophenyl)-1-propene
Hazard: Irritant-Harmful
Formula:
C₉H₉Cl MW: 152.63 g/mol |
MDL Number:
MFCD01319545 CAS Number: 3840-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...