4-(2-Chlorophenyl)-1-butene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(But-3-en-1-yl)chlorobenzene

086185-1G 086185-2G 086185-5G
102847-188EA 592.31 USD
102847-188 102847-190 102847-192
4-(2-Chlorophenyl)-1-butene ≥97%
4-(2-Chlorophenyl)-1-butene

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₁₀H₁₁Cl
MW: 166.65 g/mol
MDL Number: MFCD01319544
CAS Number: 3047-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR