3-(2-Chlorophenyl)-1-propene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-(2-propen-1-yl)benzene, 1-Allyl-2-chlorobenzene, 1-Chloro-2-(prop-2-en-1-yl)benzene, 3-(2-Chlorophenyl)prop-1-ene

086184-1G 086184-2G 086184-5G
102847-182EA 387.69 USD
102847-182 102847-184 102847-186
3-(2-Chlorophenyl)-1-propene ≥97%
3-(2-Chlorophenyl)-1-propene

Hazard: Irritant-Harmful/Light Sensitive

Formula: C₉H₉Cl
MW: 152.63 g/mol
MDL Number: MFCD00135025
CAS Number: 1587-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR