4-(4-Bromophenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086183-1G 086183-2G 086183-5G
102847-176EA 946.15 USD
102847-176 102847-178 102847-180
4-(4-Bromophenyl)-1-butene ≥97%
4-(4-Bromophenyl)-1-butene

Hazard: Irritant/Light Sensitive

Formula: C₁₀H₁₁Br
MDL Number: MFCD01319543
CAS Number: 15451-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR