3-(4-Bromophenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086182-1G 086182-2G 086182-5G
102847-170EA 646.15 USD
102847-170 102847-172 102847-174
3-(4-Bromophenyl)-1-propene ≥97%
3-(4-Bromophenyl)-1-propene

Hazard: Irritant/Light Sensitive

Formula: C₉H₉Br
MDL Number: MFCD01319542
CAS Number: 2294-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR