3-(3-Bromophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Allyl-3-bromobenzene
086181-1G
086181-2G
086181-5G
102847-164EA
620
USD
102847-164
102847-166
102847-168
3-(3-Bromophenyl)-1-propene ≥97%
3-(3-Bromophenyl)-1-propene
Hazard: Irritant-Harmful/Light Sensitive
Formula:
C₉H₉Br MW: 197.08 g/mol Boiling Pt: 45…47 °C (0.05 mmHg) |
MDL Number:
MFCD01319540 CAS Number: 18257-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...