4-(2-Bromophenyl)-1-butene ≥97%
Supplier: Matrix Scientific
086180-1G
086180-2G
086180-5G
102847-158EA
946.15
USD
102847-158
102847-160
102847-162
4-(2-Bromophenyl)-1-butene ≥97%
4-(2-Bromophenyl)-1-butene
Hazard: Irritant
Formula:
C₁₀H₁₁Br |
MDL Number:
MFCD01319539 CAS Number: 71813-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...