2-Bromo-1-undecene ≥97%

Supplier: Matrix Scientific

086179-1G 086179-2G 086179-5G
102847-152EA 592.31 USD
102847-152 102847-154 102847-156
2-Bromo-1-undecene ≥97%
2-Bromo-1-undecene

Hazard: Irritant

Formula: C₁₁H₂₁Br
MDL Number: MFCD00671863
CAS Number: 145732-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR