2-Bromo-1-octene ≥95%
Supplier: Matrix Scientific
086178-1G
086178-2G
086178-5G
102847-146EA
592.31
USD
102847-146
102847-148
102847-150
2-Bromo-1-octene ≥95%
2-Bromo-1-octene
Hazard: Irritant
Formula:
C₈H₁₅Br |
MDL Number:
MFCD00671860 CAS Number: 13249-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...