Ethyl-7-octenoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 7-octenoate
086173-1G
086173-2G
086173-5G
102847-116EA
583.08
USD
102847-116
102847-118
102847-120
Ethyl-7-octenoate ≥97%
Ethyl-7-octenoate
Hazard: Irritant
Formula:
C₁₀H₁₈O₂ |
MDL Number:
MFCD00671846 CAS Number: 35194-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...