Ethyl-5-hexenoate ≥97%

Supplier: Matrix Scientific

086172-1G 086172-2G 086172-5G
102847-110EA 387.69 USD
102847-110 102847-112 102847-114
Ethyl-5-hexenoate ≥97%
Ethyl-5-hexenoate

Hazard: Irritant

Formula: C₈H₁₄O₂
MDL Number: MFCD00671844
CAS Number: 54653-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR