7-Chloro-2-methyl-1-heptene ≥97%

Supplier: Matrix Scientific

086171-1G 086171-2G 086171-5G
102847-104EA 387.69 USD
102847-104 102847-106 102847-108
7-Chloro-2-methyl-1-heptene ≥97%
7-Chloro-2-methyl-1-heptene

Hazard: Irritant

Formula: C₈H₁₅Cl
MDL Number: MFCD00671835
CAS Number: 191488-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR