9-Chloro-1-nonene ≥97%

Supplier: Matrix Scientific

Synonyms: 9-Chloronon-1-ene

086170-1G 086170-2G 086170-5G
102847-098EA 387.69 USD
102847-098 102847-100 102847-102
9-Chloro-1-nonene ≥97%
9-Chloro-1-nonene

Hazard: Irritant

Formula: C₉H₁₇Cl
MW: 160.69 g/mol
MDL Number: MFCD00671828
CAS Number: 872-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR