7-Chloro-1-heptene ≥97%

Supplier: Matrix Scientific

086169-1G 086169-5G
102847-092EA 135.38 USD
102847-092 102847-096
7-Chloro-1-heptene ≥97%
7-Chloro-1-heptene

Hazard: Irritant

Formula: C₇H₁₃Cl
MDL Number: MFCD00671826
CAS Number: 929-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR