3-Chloro-2,5,6-trifluoropyridine ≥98%

Supplier: Matrix Scientific

084078-1G 084078-5G 084078-25G
102841-470EA 44.62 USD
102841-470 102841-472 102841-474
3-Chloro-2,5,6-trifluoropyridine ≥98%
3-Chloro-2,5,6-trifluoropyridine

Hazard: Corrosive

Formula: C₅HClF₃N
Boiling Pt: 132…134 °C
MDL Number: MFCD03001160
CAS Number: 2879-42-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR